Assay Details
| Gene Symbol: Sms | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009214.3 | NP_033240.3 | 7 - 8 | 838 | 92 | |
| GenBank mRNA | AF031486.1 | - | 7 - 8 | 821 | 92 | |
| AK017775.1 | - | 7 - 8 | 809 | 92 | ||
| AK049787.1 | - | 7 - 8 | 92 | |||
| AK136798.1 | - | 7 - 8 | 839 | 92 | ||
| AK149929.1 | - | 7 - 8 | 836 | 92 | ||
| AK160659.1 | - | 7 - 8 | 839 | 92 | ||
| AK166665.1 | - | 7 - 8 | 864 | 92 | ||
| BC046623.1 | - | 7 - 8 | 820 | 6315420 | 92 | |
| BC058688.1 | - | 7 - 8 | 1079 | 6413869 | 92 | |
| Y09419.1 | - | 7 - 8 | 811 | 92 | ||
More Information
Set Membership: |
|

Sms
Genomic Map