Assay Details
| Gene Symbol: Dsg1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181682.1 | NP_859010.1 | 11 - 12 | 1843 | 127 | |
| GenBank mRNA | AK036986.1 | - | 11 - 12 | 1951 | 127 | |
| AY192158.1 | - | 11 - 12 | 1843 | 127 | ||
| AY315940.1 | - | 1743 | 127 | |||
| BC137927.1 | - | 11 - 12 | 1834 | 8860946 | 127 | |
More Information
Set Membership: |
|

Dsg1b
Genomic Map