Assay Details
| Gene Symbol: Prpmp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024705.2 | NP_001019876.2 | 3 | 918 | 97 | |
| GenBank mRNA | AK132583.1 | - | 4 | 548 | 97 | |
| BC055856.1 | - | 3 | 909 | 4910526 | 97 | |
| BC061494.1 | - | 3 | 909 | 97 | ||
| BC080748.1 | - | 3 | 909 | 97 | ||
| BC092075.1 | - | 3 | 913 | 4920522 | 97 | |
| BC110664.1 | - | 3 | 916 | 4191067 | 97 | |
| X63004.1 | - | 811 | 93 | |||
More Information
Set Membership: |
|

Prpmp5
Genomic Map