Assay Details
| Gene Symbol: Cxcl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009140.2 | NP_033166.1 | 3 | 314 | 109 | |
| GenBank mRNA | AK137628.1 | - | 3 | 342 | 109 | |
| AK150450.1 | - | 3 | 222 | 109 | ||
| AK155916.1 | - | 3 | 341 | 109 | ||
| AK157079.1 | - | 3 | 341 | 109 | ||
| BC119511.1 | - | 3 | 288 | 40055368 | 109 | |
| BC119512.1 | - | 3 | 288 | 40055369 | 109 | |
| X53798.1 | - | 3 | 314 | 109 | ||
More Information
Set Membership: |
|

Cxcl2
Genomic Map