Assay Details
| Gene Symbol: Acat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009338.3 | NP_033364.2 | 9 | 1817 | 70 | |
| GenBank mRNA | AK037249.1 | - | 9 | 1804 | 70 | |
| AK151751.1 | - | 9 | 1805 | 70 | ||
| AK166813.1 | - | 9 | 1828 | 70 | ||
| AK167234.1 | - | 9 | 1817 | 70 | ||
| BC004823.1 | - | 9 | 1778 | 2598868 | 70 | |
| BC012496.1 | - | 9 | 1793 | 4500929 | 70 | |
More Information
Set Membership: |
|

Acat2
Genomic Map