Assay Details
| Gene Symbol: Ptp4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011200.2 | NP_035330.1 | 6 | 3024 | 89 | |
| GenBank mRNA | AK078120.1 | - | 1 | 1331 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptp4a1| Gene Symbol: Ptp4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011200.2 | NP_035330.1 | 6 | 3024 | 89 | |
| GenBank mRNA | AK078120.1 | - | 1 | 1331 | 89 | |
Set Membership: |
|