Assay Details
| Gene Symbol: Prps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021463.4 | NP_067438.1 | 7 | 1607 | 97 | |
| GenBank mRNA | AB025048.1 | - | 7 | 1546 | 97 | |
| AK089009.1 | - | 7 | 1602 | 97 | ||
| AK143609.1 | - | 7 | 1585 | 97 | ||
| AK165524.1 | - | 7 | 1586 | 97 | ||
| AK165987.1 | - | 7 | 1588 | 97 | ||
| AK166856.1 | - | 7 | 1586 | 97 | ||
| AK168036.1 | - | 7 | 1519 | 97 | ||
| BC054772.1 | - | 7 | 1558 | 6415720 | 97 | |
More Information
Set Membership: |
|

Prps1
Genomic Map