Assay Details
| Gene Symbol: Zfp935 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136496.1 | NP_001129968.1 | 3 | 1857 | 78 | |
| NM_178875.4 | NP_849206.2 | 4 | 1918 | 78 | ||
| GenBank mRNA | AK018441.1 | - | 3 | 1848 | 78 | |
| AY189975.1 | - | 3 | 1834 | 78 | ||
| BC012524.1 | - | 3 | 1833 | 4008347 | 78 | |
| BC047396.1 | - | 4 | 1895 | 3979342 | 78 | |
| BC087936.1 | - | 4 | 1918 | 78 | ||
| BC113193.1 | - | 4 | 1914 | 40062446 | 78 | |
More Information
Set Membership: |
|

Zfp935
Genomic Map