Assay Details
| Gene Symbol: Ccl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011329.3 | NP_035459.1 | 1 - 2 | 149 | 65 | |
| GenBank mRNA | AF065924.1 | - | 1 - 2 | 128 | 65 | |
| AF065925.1 | - | 1 - 2 | 128 | 65 | ||
| AF065926.1 | - | 1 - 2 | 128 | 65 | ||
| AK019040.1 | - | 1 - 2 | 149 | 65 | ||
| BC120806.1 | - | 1 - 2 | 140 | 40129729 | 65 | |
| M17957.1 | - | 1 - 2 | 152 | 65 | ||
| M23501.1 | - | 1 - 2 | 149 | 65 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2157829 | M23501.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1960386 | BC042744.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1960386 | M17957.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1960386 | M23501.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1960386 | NM_011329.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1960386 | AK019040.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 404373 | AK087064.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 88503 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P139353 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1960386 | BC042744.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1960386 | M17957.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1960386 | M23501.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1960386 | NM_011329.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1960386 | AK019040.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P299896 | BC042744.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P299896 | BC120806.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P299896 | M17957.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P299896 | M23501.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P299896 | NM_011329.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964630 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964636 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964658 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964660 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964626 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964611 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964624 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964634 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964600 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964598 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964617 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964638 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964605 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964613 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964619 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964656 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964649 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964604 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964655 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964625 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964646 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964606 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964632 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964647 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964643 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964662 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964650 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964641 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964642 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964610 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964645 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964653 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964599 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964622 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964639 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964608 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964616 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964602 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P139353 | - | - |
| Whole Mouse Genome G4122A | A_51_P299896 | BC042744.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P299896 | BC120806.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P299896 | M17957.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P299896 | M23501.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P299896 | NM_011329.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134669 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P139353 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09436305 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09152164 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09436303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964622 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964608 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964619 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964598 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964566 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09269093 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09332616 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964617 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964605 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964636 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964622 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09152202 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09269090 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964670 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964582 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964602 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964641 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964534 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09287030 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964568 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09332631 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964688 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08983165 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09350593 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964639 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964565 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964632 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09332628 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964634 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964596 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09152188 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964606 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964695 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964588 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09152164 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964584 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964562 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964577 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964610 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964591 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964669 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09152204 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964612 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964650 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964690 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964660 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964696 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964604 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09152199 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09560838 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964592 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964697 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964613 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09152190 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09614965 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09523322 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964627 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964667 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964642 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964575 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964599 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09152187 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964666 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964658 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09152183 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964629 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964593 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964586 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964574 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964638 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09152205 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964673 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964678 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964699 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964663 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10389067 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10389066 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10389069 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10389068 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10389065 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1421688_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10389064 | - | - |
| Mouse Genome 430 2.0 Array | 1421688_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1421688_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20290_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20290_PM_at | - | - |
| Affymetrix Murine Genome 6500A Array | Msa11890_at | - | - |
| Mouse Expression Set 430 | 1421688_a_at | - | - |
| Mouse Expression Set 430 | 1421688_PM_a_at | - | - |
| Murine Genome U74v2 Set | 94166_PM_g_at | - | - |
| Murine Genome U74v2 Set | 94165_PM_at | - | - |
| Murine Genome U74v2 Set | 94165_at | - | - |
| Murine Genome U74v2 Set | 94166_g_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421688_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421688_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421688_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421688_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4230167 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004230167 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1255735 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1255735 | - | - |
| mouse-6 v1.1 (Illumina) | scl020290.3_11-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4230167 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl020290.3_11-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4230167 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1255735 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1255735 | - | - |

Ccl1
Genomic Map