Assay Details
| Gene Symbol: SEV-KOS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | SENDAI_KLF4_KOS.0 | - | 401 | 80 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SEV-KOS| Gene Symbol: SEV-KOS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | SENDAI_KLF4_KOS.0 | - | 401 | 80 | ||
Set Membership: |
|