Assay Details
| Gene Symbol: mStrawberry | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY678266.1 | - | 302 | 51 | ||
| Gene Symbol: E2-Crimson | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | HE583184.1 | - | 287 | 51 | ||
| Gene Symbol: mBanana | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | mBanana_Clontech.0 | - | 302 | 51 | ||
| Gene Symbol: mCherry | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | MK160997.1 | - | 302 | 51 | ||
| Gene Symbol: mPlum | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY786537.1 | - | 290 | 51 | ||
| Gene Symbol: mRasberry | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY786536.1 | - | 290 | 51 | ||
| Gene Symbol: tdTomato | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | KT878736.1 | - | 290 | 51 | ||
More Information
Set Membership: |
|

mStrawberry
Genomic Map