Assay Details
| Gene Symbol: PPP4C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027962053.1 | XP_027817854.1 | 8 - 9 | 969 | 54 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PPP4C| Gene Symbol: PPP4C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027962053.1 | XP_027817854.1 | 8 - 9 | 969 | 54 | |
Set Membership: |
|