Assay Details
| Gene Symbol: UBE2Q2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004017823.4 | XP_004017872.2 | 6 - 7 | 793 | 99 | |
| XM_004017824.4 | XP_004017873.2 | 5 - 6 | 679 | 99 | ||
| XM_012098644.3 | XP_011954034.2 | 6 - 7 | 753 | 99 | ||
More Information
Set Membership: |
|

UBE2Q2
Genomic Map