Assay Details
| Gene Symbol: ARHGEF9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004022026.4 | XP_004022075.1 | 9 - 10 | 2207 | 76 | |
| XM_012106570.3 | XP_011961960.1 | 9 - 10 | 2203 | 76 | ||
| XM_012106574.3 | XP_011961964.1 | 9 - 10 | 2196 | 76 | ||
| XM_012106575.2 | XP_011961965.1 | 9 - 10 | 2292 | 76 | ||
| XM_015104868.2 | XP_014960354.1 | 11 - 12 | 1900 | 76 | ||
| XM_027963058.1 | XP_027818859.1 | 11 - 12 | 3166 | 76 | ||
| XM_027963059.1 | XP_027818860.1 | 10 - 11 | 2343 | 76 | ||
| XM_027963061.1 | XP_027818862.1 | 10 - 11 | 2366 | 76 | ||
| XM_027963062.1 | XP_027818863.1 | 8 - 9 | 2027 | 76 | ||
More Information
Set Membership: |
|

ARHGEF9
Genomic Map