Assay Details
| Gene Symbol: DCUN1D5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015100872.2 | XP_014956358.2 | 2 - 3 | 453 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DCUN1D5| Gene Symbol: DCUN1D5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015100872.2 | XP_014956358.2 | 2 - 3 | 453 | 61 | |
Set Membership: |
|