Assay Details
| Gene Symbol: PPP1CB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027966639.1 | XP_027822440.1 | 5 - 6 | 881 | 65 | |
| XM_027966640.1 | XP_027822441.1 | 6 - 7 | 765 | 65 | ||
| XM_027966641.1 | XP_027822442.1 | 6 - 7 | 744 | 65 | ||
| XM_027966642.1 | XP_027822443.1 | 6 - 7 | 801 | 65 | ||
More Information
Set Membership: |
|

PPP1CB
Genomic Map