Assay Details
| Gene Symbol: IST1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004015113.4 | XP_004015162.1 | 4 - 5 | 484 | 69 | |
| XM_012190049.3 | XP_012045439.1 | 4 - 5 | 484 | 69 | ||
| XM_012190050.3 | XP_012045440.1 | 4 - 5 | 488 | 69 | ||
More Information
Set Membership: |
|

IST1
Genomic Map