Assay Details
| Gene Symbol: QPCT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166178.1 | NP_001159650.1 | 2 - 3 | 269 | 68 | |
| GenBank mRNA | AF039441.1 | - | 1 - 2 | 55 | 68 | |
| GQ221058.1 | - | 2 - 3 | 269 | 68 | ||
More Information
Set Membership: |
|

QPCT
Genomic Map