Assay Details
| Gene Symbol: LMAN2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004006157.4 | XP_004006206.2 | 6 - 7 | 800 | 65 | |
| XM_004006158.3 | XP_004006207.2 | 7 - 8 | 837 | 65 | ||
| XM_027967094.1 | XP_027822895.1 | 7 - 8 | 2350 | 65 | ||
| XM_027967095.1 | XP_027822896.1 | 4 - 5 | 1788 | 65 | ||
| XM_027967096.1 | XP_027822897.1 | 6 - 7 | 2020 | 65 | ||
| XM_027967097.1 | XP_027822898.1 | 5 - 6 | 1869 | 65 | ||
More Information
Set Membership: |
|

LMAN2L
Genomic Map