Assay Details
| Gene Symbol: EMP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004015346.4 | XP_004015395.1 | 4 - 5 | 620 | 63 | |
| XM_004015347.3 | XP_004015396.1 | 4 - 5 | 744 | 63 | ||
| XM_012190615.3 | XP_012046005.1 | 4 - 5 | 446 | 63 | ||
More Information
Set Membership: |
|

EMP3
Genomic Map