Assay Details
| Gene Symbol: NFIC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027969890.1 | XP_027825691.1 | 3 - 4 | 882 | 61 | |
| XM_027969892.1 | XP_027825693.1 | 3 - 4 | 674 | 61 | ||
| XM_027969893.1 | XP_027825694.1 | 3 - 4 | 859 | 61 | ||
| XM_027969894.1 | XP_027825695.1 | 3 - 4 | 774 | 61 | ||
| XM_027969895.1 | XP_027825696.1 | 3 - 4 | 753 | 61 | ||
| XM_027969896.1 | XP_027825697.1 | 3 - 4 | 674 | 61 | ||
More Information
Set Membership: |
|

NFIC
Genomic Map