Assay Details
| Gene Symbol: EIF3G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004008511.3 | XP_004008560.1 | 2 - 3 | 147 | 54 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EIF3G| Gene Symbol: EIF3G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004008511.3 | XP_004008560.1 | 2 - 3 | 147 | 54 | |
Set Membership: |
|