Assay Details
| Gene Symbol: GUCA1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027958443.1 | XP_027814244.1 | 2 - 3 | 1183 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GUCA1A| Gene Symbol: GUCA1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027958443.1 | XP_027814244.1 | 2 - 3 | 1183 | 66 | |
Set Membership: |
|