Assay Details
| Gene Symbol: C1QBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004012589.4 | XP_004012638.2 | 4 - 5 | 687 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C1QBP| Gene Symbol: C1QBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004012589.4 | XP_004012638.2 | 4 - 5 | 687 | 65 | |
Set Membership: |
|