Assay Details
| Gene Symbol: SNAI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126342.1 | NP_001119814.1 | 2 - 3 | 619 | 72 | |
| GenBank mRNA | EU583469.1 | - | 2 - 3 | 619 | 72 | |
More Information
Set Membership: |
|

SNAI2
Genomic Map