Assay Details
| Gene Symbol: MRPL24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002631.4 | XP_004002680.1 | 2 - 3 | 328 | 67 | |
| XM_012183478.3 | XP_012038868.1 | 2 - 3 | 331 | 67 | ||
| XM_015092309.2 | XP_014947795.1 | 2 - 3 | 266 | 67 | ||
| XM_027976525.1 | XP_027832326.1 | 2 - 3 | 784 | 67 | ||
More Information
Set Membership: |
|

MRPL24
Genomic Map