Assay Details
| Gene Symbol: PSMA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004007997.4 | XP_004008046.1 | 5 - 6 | 512 | 71 | |
| GenBank mRNA | JX534539.1 | - | 1 | 453 | 71 | |
More Information
Set Membership: |
|

PSMA2
Genomic Map