Assay Details
Gene Symbol: MORN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004017347.4 | XP_004017396.1 | 2 - 3 | 760 | 69 | |
XM_027956477.1 | XP_027812278.1 | 2 - 3 | 760 | 69 | ||
XM_027956478.1 | XP_027812279.1 | 3 - 4 | 454 | 69 | ||
XM_027956479.1 | XP_027812280.1 | 3 - 4 | 436 | 69 | ||
XM_027956480.1 | XP_027812281.1 | 2 - 3 | 733 | 69 | ||
XM_027956482.1 | XP_027812283.1 | 2 - 3 | 545 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
