Assay Details
| Gene Symbol: TSKS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027978637.1 | XP_027834438.1 | 6 - 7 | 1207 | 76 | |
| XM_027978638.1 | XP_027834439.1 | 6 - 7 | 1180 | 76 | ||
| XM_027978639.1 | XP_027834440.1 | 6 - 7 | 1207 | 76 | ||
| XM_027978640.1 | XP_027834441.1 | 6 - 7 | 1180 | 76 | ||
| XM_027978641.1 | XP_027834442.1 | 6 - 7 | 1207 | 76 | ||
| XM_027978642.1 | XP_027834443.1 | 6 - 7 | 1180 | 76 | ||
| XM_027978643.1 | XP_027834444.1 | 2 - 3 | 673 | 76 | ||
| XM_027978644.1 | XP_027834445.1 | 6 - 7 | 1207 | 76 | ||
| XM_027978645.1 | XP_027834446.1 | 6 - 7 | 1180 | 76 | ||
More Information
Set Membership: |
|

TSKS
Genomic Map