Assay Details
| Gene Symbol: CCDC58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012095631.2 | XP_011951021.2 | 1 - 2 | 98 | 76 | |
| XM_012095634.2 | XP_011951024.2 | 1 - 2 | 112 | 76 | ||
More Information
Set Membership: |
|

CCDC58
Genomic Map