Assay Details
| Gene Symbol: ALG5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012184556.3 | XP_012039946.1 | 4 - 5 | 324 | 69 | |
| XM_012184557.3 | XP_012039947.1 | 4 - 5 | 423 | 69 | ||
| XM_027973605.1 | XP_027829406.1 | 4 - 5 | 385 | 69 | ||
More Information
Set Membership: |
|

ALG5
Genomic Map