Assay Details
| Gene Symbol: CDKN2AIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015104556.2 | XP_014960042.2 | 1 - 2 | 454 | 79 | |
| XM_015104557.2 | XP_014960043.2 | 1 - 2 | 457 | 79 | ||
More Information
Set Membership: |
|

CDKN2AIP
Genomic Map