Assay Details
| Gene Symbol: SEPTIN10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015094375.2 | XP_014949861.2 | 6 - 7 | 811 | 106 | |
| XM_015094376.2 | XP_014949862.2 | 6 - 7 | 822 | 106 | ||
More Information
Set Membership: |
|

SEPTIN10
Genomic Map