Assay Details
| Gene Symbol: PLPPR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027969684.1 | XP_027825485.1 | 6 - 7 | 981 | 68 | |
| XM_027969686.1 | XP_027825487.1 | 8 - 9 | 1299 | 68 | ||
| XM_027969687.1 | XP_027825488.1 | 5 - 6 | 791 | 68 | ||
More Information
Set Membership: |
|

PLPPR2
Genomic Map