Assay Details
| Gene Symbol: LOC114118746 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027980144.1 | XP_027835945.1 | 1 | 1227 | 103 | |
| Gene Symbol: SUCLA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973559.1 | XP_027829360.1 | 9 - 10 | 1257 | 103 | |
More Information
Set Membership: |
|

LOC114118746
Genomic Map