Assay Details
| Gene Symbol: LOC114117592 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027976874.1 | XP_027832675.1 | 4 - 5 | 838 | 106 | |
| XM_027976875.1 | XP_027832676.1 | 4 - 5 | 840 | 106 | ||
| XM_027976876.1 | XP_027832677.1 | 4 - 5 | 840 | 106 | ||
| Gene Symbol: LOC101103001 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027976879.1 | XP_027832680.1 | 4 - 5 | 837 | 106 | |
| Gene Symbol: PCED1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027976374.1 | XP_027832175.1 | 4 - 5 | 567 | 106 | |
More Information
Set Membership: |
|

LOC114117592
Genomic Map