Assay Details
| Gene Symbol: ETV6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027967861.1 | XP_027823662.1 | 7 - 8 | 1397 | 68 | |
| XM_027967863.1 | XP_027823664.1 | 7 - 8 | 1394 | 68 | ||
| XM_027967864.1 | XP_027823665.1 | 7 - 8 | 3109 | 68 | ||
More Information
Set Membership: |
|

ETV6
Genomic Map