Assay Details
| Gene Symbol: ZDHHC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004018524.4 | XP_004018573.1 | 4 - 5 | 787 | 66 | |
| XM_012100193.3 | XP_011955583.1 | 4 - 5 | 774 | 66 | ||
| XM_027957745.1 | XP_027813546.1 | 4 - 5 | 774 | 66 | ||
| XM_027957746.1 | XP_027813547.1 | 4 - 5 | 783 | 66 | ||
| XM_027957747.1 | XP_027813548.1 | 4 - 5 | 774 | 66 | ||
More Information
Set Membership: |
|

ZDHHC3
Genomic Map