Assay Details
| Gene Symbol: SYPL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015092157.2 | XP_014947643.2 | 5 - 6 | 883 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SYPL2| Gene Symbol: SYPL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015092157.2 | XP_014947643.2 | 5 - 6 | 883 | 69 | |
Set Membership: |
|