Assay Details
| Gene Symbol: PDE12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027958256.1 | XP_027814057.1 | 2 - 3 | 1480 | 124 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PDE12| Gene Symbol: PDE12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027958256.1 | XP_027814057.1 | 2 - 3 | 1480 | 124 | |
Set Membership: |
|