Assay Details
| Gene Symbol: GOLGA7B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004020092.3 | XP_004020141.1 | 2 - 3 | 639 | 78 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

GOLGA7B| Gene Symbol: GOLGA7B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004020092.3 | XP_004020141.1 | 2 - 3 | 639 | 78 | |