Assay Details
| Gene Symbol: ERICH5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004023595.4 | XP_004023644.2 | 2 - 3 | 1177 | 77 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

ERICH5| Gene Symbol: ERICH5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004023595.4 | XP_004023644.2 | 2 - 3 | 1177 | 77 | |