Assay Details
| Gene Symbol: CORO1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027956652.1 | XP_027812453.1 | 4 - 5 | 579 | 56 | |
| XM_027956653.1 | XP_027812454.1 | 4 - 5 | 553 | 56 | ||
More Information
Set Membership: |
|

CORO1C
Genomic Map