Assay Details
| Gene Symbol: BMPR1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009431.1 | NP_001009431.1 | 9 - 10 | 1229 | 65 | |
| XM_027970721.1 | XP_027826522.1 | 8 - 9 | 1503 | 65 | ||
| XM_027970722.1 | XP_027826523.1 | 8 - 9 | 1371 | 65 | ||
| XM_027970723.1 | XP_027826524.1 | 10 - 11 | 1416 | 65 | ||
| XM_027970724.1 | XP_027826525.1 | 10 - 11 | 1411 | 65 | ||
| XM_027970725.1 | XP_027826526.1 | 8 - 9 | 1167 | 65 | ||
| XM_027970726.1 | XP_027826527.1 | 8 - 9 | 2165 | 65 | ||
| GenBank mRNA | AF298885.1 | - | 8 - 9 | 1120 | 65 | |
| AF312016.1 | - | 8 - 9 | 1157 | 65 | ||
| AF357007.1 | - | 9 - 10 | 1229 | 65 | ||
| EU183347.1 | - | 8 - 9 | 1142 | 65 | ||
More Information
Set Membership: |
|

BMPR1B
Genomic Map