Assay Details
| Gene Symbol: POLDIP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004007028.3 | XP_004007077.1 | 6 - 7 | 979 | 68 | |
| XM_004007029.3 | XP_004007078.1 | 5 - 6 | 892 | 68 | ||
| XM_012175731.2 | XP_012031121.1 | 5 - 6 | 802 | 68 | ||
More Information
Set Membership: |
|

POLDIP3
Genomic Map