Assay Details
| Gene Symbol: SESN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004011221.3 | XP_004011270.1 | 2 - 3 | 1120 | 60 | |
| XM_004011222.3 | XP_004011271.2 | 2 - 3 | 278 | 60 | ||
| XM_012182656.2 | XP_012038046.1 | 2 - 3 | 1120 | 60 | ||
| XM_012182657.2 | XP_012038047.2 | 2 - 3 | 265 | 60 | ||
More Information
Set Membership: |
|

SESN1
Genomic Map