Assay Details
| Gene Symbol: ISOC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012178174.3 | XP_012033564.2 | 4 - 5 | 771 | 80 | |
| XM_027969967.1 | XP_027825768.1 | 4 - 5 | 744 | 80 | ||
More Information
Set Membership: |
|

ISOC1
Genomic Map