Assay Details
| Gene Symbol: MCUR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027958368.1 | XP_027814169.1 | 7 - 8 | 1022 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MCUR1| Gene Symbol: MCUR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027958368.1 | XP_027814169.1 | 7 - 8 | 1022 | 69 | |
Set Membership: |
|