Assay Details
| Gene Symbol: TCP11L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004006693.4 | XP_004006742.1 | 4 - 5 | 588 | 72 | |
| XM_012174949.3 | XP_012030339.1 | 3 - 4 | 398 | 72 | ||
More Information
Set Membership: |
|

TCP11L2
Genomic Map