Assay Details
| Gene Symbol: LOC101112585 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004013721.4 | XP_004013770.1 | 4 - 5 | 875 | 108 | |
| XM_012187328.2 | XP_012042718.1 | 3 - 4 | 498 | 108 | ||
| XM_027975640.1 | XP_027831441.1 | 3 - 4 | 939 | 108 | ||
More Information
Set Membership: |
|

LOC101112585
Genomic Map